Gene Transcripts

Gene Symbol
Cbx8
Entrez Gene ID
303731
Gene Name
chromobox homolog 8
Gene Aliases
Chromosome Location
Chr. 10: 108148073 - 108150511 on Build Rnor_6.0
UniGene ID
Rn.43722
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034078.1 NP_001029250.1 5 1048
GenBank mRNA BC099206.1 - 5 1048

View Full Product Details