Gene Transcripts

Gene Symbol
C1qtnf6
Entrez Gene ID
315114
Gene Name
C1q and tumor necrosis factor related protein 6
Gene Aliases
Ctrp6
Chromosome Location
Chr. 7: 119746150 - 119752720 on Build Rnor_6.0
UniGene ID
Rn.17708
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034932.1 NP_001030104.1 3 1935
GenBank mRNA BC101907.1 - 3 1935

View Full Product Details