Gene Transcripts

Gene Symbol
Slc11a1
Entrez Gene ID
316519
Gene Name
solute carrier family 11 (proton-coupled divalent metal ion transporter), member 1
Gene Aliases
Bcg, Itg, Lsh, Nramp1
Chromosome Location
Chr. 9: 81655610 - 81666697 on Build Rnor_6.0
UniGene ID
Rn.105919
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001031658.1 NP_001026828.1 14 1897
GenBank mRNA BC100653.1 - 14 1897

View Full Product Details