Gene Transcripts

Gene Symbol
Ifna14
Entrez Gene ID
404549
Gene Name
interferon alpha 14
Gene Aliases
OTTMUSG00000007662
Chromosome Location
Chr. 4: 88571229 - 88571798 on Build GRCm38
UniGene ID
Mm.377947
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_206975.1 NP_996858.1 1 341
GenBank mRNA BC120722.1 - 1 391
BC125321.1 - 1 391 40129836

View Full Product Details