Gene Transcripts
- Gene Symbol
- TRIM49
- Entrez Gene ID
- 57093
- Gene Name
- tripartite motif containing 49
- Gene Aliases
- RNF18, TRIM49A, TRIM49L2
- Chromosome Location
- Chr. 11: 89797655 - 89808575 on Build GRCh38
- UniGene ID
- Hs.534218
- Species
- Homo sapiens
- Gene Symbol
- TRIM49B
- Entrez Gene ID
- 283116
- Gene Name
- tripartite motif containing 49B
- Gene Aliases
- Chromosome Location
- Chr. 11: 49031596 - 49038049 on Build GRCh38
- UniGene ID
- Hs.448644
- Species
- Homo sapiens
- Gene Symbol
- TRIM49C
- Entrez Gene ID
- 642612
- Gene Name
- tripartite motif containing 49C
- Gene Aliases
- TRIM49L2
- Chromosome Location
- Chr. 11: 90031106 - 90042025 on Build GRCh38
- UniGene ID
- Hs.740080
- Species
- Homo sapiens
View Full Product Details