Gene Transcripts

Gene Symbol
Scgb2b20
Entrez Gene ID
494519
Gene Name
secretoglobin, family 2B, member 20
Gene Aliases
Abpbg20, Abpd, C2c, Scgb2b2
Chromosome Location
Chr. 7: 33364343 - 33366319 on Build GRCm38
UniGene ID
Mm.389857
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009952.2 NP_001009952.2 2 115
GenBank mRNA AF272844.1 - 2 119
CK616187.1 - 2 116

View Full Product Details