Gene Transcripts

Gene Symbol
C1qtnf2
Entrez Gene ID
69183
Gene Name
C1q and tumor necrosis factor related protein 2
Gene Aliases
1810033K05Rik, CTRP2
Chromosome Location
Chr. 11: 43474284 - 43491525 on Build GRCm38
UniGene ID
Mm.390374
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026979.5 NP_081255.1 3 221
GenBank mRNA AK007683.1 - 3 222
BC030324.1 - 3 222 5375090
DQ002395.1 - 2 83

View Full Product Details