Gene Transcripts

Gene Symbol
Atad5
Entrez Gene ID
237877
Gene Name
ATPase family, AAA domain containing 5
Gene Aliases
C130052G03Rik, FRAG1, Gm17
Chromosome Location
Chr. 11: 80089379 - 80135797 on Build GRCm38
UniGene ID
Mm.89551
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001029856.2 NP_001025027.1 2 1611
GenBank mRNA AK032814.1 - 2 1549
AK048355.1 - 2 1573
AK053857.1 - 2 1601
AK085211.1 - 2 1602
AK165945.1 - 2 1578
AY557610.1 - 2 1602
BC038279.1 - 2 1619
BC141180.1 - 2 1269 9055792

View Full Product Details