Gene Transcripts

Gene Symbol
ADPRM
Entrez Gene ID
56985
Gene Name
ADP-ribose/CDP-alcohol diphosphatase, manganese dependent
Gene Aliases
C17orf48, MDS006, NBLA03831
Chromosome Location
Chr. 17: 10697610 - 10711558 on Build GRCh38
UniGene ID
Hs.47668
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK292629.1 - 2 650
BC001294.2 - 1 639 3452502
BC070155.1 - 2 679 4656998

View Full Product Details