Gene Transcripts
- Gene Symbol
- TIFA
- Entrez Gene ID
- 92610
- Gene Name
- TRAF interacting protein with forkhead associated domain
- Gene Aliases
- T2BP, T6BP, TIFAA
- Chromosome Location
- Chr. 4: 112275626 - 112285903 on Build GRCh38
- UniGene ID
- Hs.310640
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_052864.2 |
NP_443096.1
|
2 |
3041 |
|
View Full Product Details