Gene Transcripts

Gene Symbol
AWAT1
Entrez Gene ID
158833
Gene Name
acyl-CoA wax alcohol acyltransferase 1
Gene Aliases
DGA2, DGAT2L3
Chromosome Location
Chr. X: 70233489 - 70240661 on Build GRCh38
UniGene ID
Hs.407641
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013579.2 NP_001013597.1 4 319
XM_017029295.1 XP_016884784.1 4 1485
GenBank mRNA AY947638.1 - 4 278
W68831.1 - 4 319

View Full Product Details