Gene Transcripts

Gene Symbol
BOLA3
Entrez Gene ID
388962
Gene Name
bolA family member 3
Gene Aliases
MMDS2
Chromosome Location
Chr. 2: 74135401 - 74147912 on Build GRCh38
UniGene ID
Hs.61472
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001035505.1 NP_001030582.1 3 443
NM_212552.2 NP_997717.2 4 532

View Full Product Details