Gene Transcripts

Gene Symbol
Bri3
Entrez Gene ID
304284
Gene Name
brain protein I3
Gene Aliases
Chromosome Location
Chr. 12: 12322247 - 12330929 on Build Rnor_6.0
UniGene ID
Rn.7700
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009604.1 NP_001009604.1 3 567
GenBank mRNA BC087646.1 - 3 567

View Full Product Details