Gene Transcripts

Gene Symbol
Amy1a
Entrez Gene ID
24203
Gene Name
amylase, alpha 1A (salivary)
Gene Aliases
Amy1
Chromosome Location
Chr. 2: 216428709 - 216443518 on Build Rnor_6.0
UniGene ID
Rn.116361
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001010970.1 NP_001010970.1 2 207
GenBank mRNA AB057450.1 - 2 207
BC088228.1 - 1 150 7323054
FQ219213.1 - 2 536

View Full Product Details