Gene Transcripts

Gene Symbol
Rnase1l2
Entrez Gene ID
305844
Gene Name
ribonuclease, RNase A family, 1-like 2 (pancreatic)
Gene Aliases
RGD1565230
Chromosome Location
Chr. 15: 28127147 - 28128797 on Build Rnor_6.0
UniGene ID
Rn.226680
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025116.1 NP_001020287.1 1 444
XM_006251875.1 XP_006251937.1 2 552

Gene Symbol
LOC103694853
Entrez Gene ID
103694853
Gene Name
ribonuclease pancreatic gamma-type
Gene Aliases
Chromosome Location
Chr. Un|NW_007906778: 117096 - 118746 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008773795.1 XP_008772017.1 552

View Full Product Details