Gene Transcripts

Gene Symbol
CYP3A7
Entrez Gene ID
1551
Gene Name
cytochrome P450 family 3 subfamily A member 7
Gene Aliases
CP37, CYPIIIA7, P-450(HFL33), P-450111A7, P450-HFLA
Chromosome Location
Chr. 7: 99705037 - 99735200 on Build GRCh38
UniGene ID
Hs.111944
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000765.4 NP_000756.3 13 1622
GenBank mRNA BC067436.1 - 13 1600
BX647143.1 - 10 1854
D00408.1 - 13 1518

View Full Product Details