Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001009936.2 | NP_001009936.1 | 2 | 344 | |
NM_001286840.1 | NP_001273769.1 | 2 | 215 | ||
NM_001286843.1 | NP_001273772.1 | 2 | 344 | ||
NM_015651.2 | NP_056466.1 | 2 | 344 | ||
XM_005251906.2 | XP_005251963.1 | 2 | 152 | ||
XM_011518509.2 | XP_011516811.1 | 2 | 195 | ||
XM_011518511.2 | XP_011516813.1 | 2 | 678 | ||
XM_011518515.2 | XP_011516817.1 | 2 | 679 | ||
XM_011518516.2 | XP_011516818.1 | 2 | 681 | ||
XM_017014612.1 | XP_016870101.1 | 2 | 184 | ||
XM_017014613.1 | XP_016870102.1 | 2 | 682 | ||
GenBank mRNA | AK122744.1 | - | 2 | 204 | |
BC022374.1 | - | 2 | 204 | ||
BC108663.1 | - | 2 | 311 | ||
BC125076.1 | - | 1 | 116 | 40117657 | |
BC125077.1 | - | 1 | 116 | 40117658 | |
BM784783.1 | - | 2 | 344 | ||
BM784790.1 | - | 2 | 344 | ||
BX640713.1 | - | 2 | 216 |