Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001127457.2 | NP_001120929.1 | 12 | 3968 | |
NM_021117.3 | NP_066940.2 | 12 | 4104 | ||
GenBank mRNA | AB014558.1 | - | 12 | 4008 | |
AK001194.1 | - | 3 | 2434 | ||
BC035161.1 | - | 15 | 4288 | 5265444 | |
BC041814.1 | - | 12 | 4032 | 5266781 | |
BG260501.1 | - | 1 | 516 | ||
EU219622.1 | - | 12 | 4027 |