Gene Transcripts

Gene Symbol
MAPKAP1
Entrez Gene ID
79109
Gene Name
mitogen-activated protein kinase associated protein 1
Gene Aliases
JC310, MIP1, SIN1, SIN1b, SIN1g
Chromosome Location
Chr. 9: 125437394 - 125707234 on Build GRCh38
UniGene ID
Hs.495138
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001006617.1 NP_001006618.1 6 1097
NM_001006618.1 NP_001006619.1 6 1097
NM_001006619.1 NP_001006620.1 6 1097
NM_001006620.1 NP_001006621.1 5 769
NM_001006621.1 NP_001006622.1 4 679
NM_024117.3 NP_077022.1 6 1097
XM_011519005.1 XP_011517307.1 7 1238
XM_011519006.2 XP_011517308.1 7 1238
XM_011519007.1 XP_011517309.1 7 1238
XM_011519008.1 XP_011517310.1 7 1238
XM_011519009.1 XP_011517311.1 7 1238
XM_011519010.2 XP_011517312.1 3 415
XM_017015126.1 XP_016870615.1 6 986
XM_017015127.1 XP_016870616.1 6 986
GenBank mRNA AK290060.1 - 6 910
AK295364.1 - 3 403
AK309507.1 - 3 364
AK310361.1 - 3 403
AK316149.1 - 3 342
AL833042.1 - 5 730
AY524429.1 - 6 1094
AY524430.1 - 6 1094
AY524431.1 - 6 1094
AY524432.1 - 6 1094
AY633624.1 - 6 1064
AY633625.1 - 6 1071
AY633626.1 - 6 1071
BC002326.2 - 6 1066 2823015
BC003044.1 - 6 1066 2823015
M37191.1 - 5 542

View Full Product Details