Gene Transcripts

Gene Symbol
CELF5
Entrez Gene ID
60680
Gene Name
CUGBP, Elav-like family member 5
Gene Aliases
BRUNOL-5, BRUNOL5, CELF-5
Chromosome Location
Chr. 19: 3224676 - 3297076 on Build GRCh38
UniGene ID
Hs.655747
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001172673.1 NP_001166144.1 4, 5 567
NM_021938.3 NP_068757.2 4, 5 568
NR_033342.1 4, 5 406
XM_006722832.1 XP_006722895.1 4, 5 595
XM_006722833.1 XP_006722896.1 4, 5 592
XM_006722836.2 XP_006722899.1 3, 4 236
XM_006722837.3 XP_006722900.1 4, 5 507
XM_006722838.3 XP_006722901.1 3, 4 379
XM_011528173.1 XP_011526475.1 3, 4 208
XM_011528174.2 XP_011526476.1 3, 4 377
XM_017027111.1 XP_016882600.1 2, 3 451
GenBank mRNA AB209067.1 - 4, 5 567
AF329266.1 - 4, 5 531
AK294109.1 - 4, 5 406
AL832550.1 - 2, 3 156
BC028101.1 - 4, 5 568 5242909

View Full Product Details