Gene Transcripts

Gene Symbol
SDF2L1
Entrez Gene ID
23753
Gene Name
stromal cell derived factor 2 like 1
Gene Aliases
Chromosome Location
Chr. 22: 21642253 - 21644299 on Build GRCh38
UniGene ID
Hs.303116
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022044.2 NP_071327.2 3 705
GenBank mRNA AB043007.1 - 3 641
AF277316.1 - 3 629
AY359118.1 - 3 672
BC132849.1 - 3 697 40146871
BC132851.1 - 3 697 40146873
CR456570.1 - 3 630

View Full Product Details