Gene Transcripts

Gene Symbol
GID4
Entrez Gene ID
79018
Gene Name
GID complex subunit 4 homolog
Gene Aliases
C17orf39, VID24
Chromosome Location
Chr. 17: 18039297 - 18068405 on Build GRCh38
UniGene ID
Hs.187422
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024052.4 NP_076957.3 6 1834
XM_005256800.1 XP_005256857.1 5 1590
GenBank mRNA AK074284.1 - 6 1709
AK127580.1 - 6 1521

View Full Product Details