Gene Transcripts

Gene Symbol
WDR77
Entrez Gene ID
79084
Gene Name
WD repeat domain 77
Gene Aliases
HKMT1069, MEP-50, MEP50, Nbla10071, p44, p44/Mep50
Chromosome Location
Chr. 1: 111439890 - 111449308 on Build GRCh38
UniGene ID
Hs.204773
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317062.1 NP_001303991.1 4 597
NM_001317063.1 NP_001303992.1 4 597
NM_024102.3 NP_077007.1 4 597
NR_133654.1 3 455
GenBank mRNA AB073603.1 - 4 545
AF052183.1 - 4 483
AF478464.1 - 4 458
AK022860.1 - 4 527
AK223189.1 - 4 527
AK296163.1 - 3 399
AL557974.3 - 4 513
AY225316.1 - 4 483
BC001679.2 - 4 518 2822173
BC006477.1 - 4 518 2822173
BC009411.1 - 4 530 3614223
BC011778.2 - 4 537 3941878
BC016946.1 - 4 497 3845908
BI195029.1 - 3 269

View Full Product Details