Gene Transcripts
- Gene Symbol
- RNF219
- Entrez Gene ID
- 79596
- Gene Name
- ring finger protein 219
- Gene Aliases
- C13orf7
- Chromosome Location
- Chr. 13: 78614286 - 78659179 on Build GRCh38
- UniGene ID
- Hs.567576
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_024546.3 |
NP_078822.3
|
3, 4 |
367 |
|
XM_011535225.1 |
XP_011533527.1
|
2, 3 |
217 |
|
XM_017020743.1 |
XP_016876232.1
|
3, 4 |
2162 |
|
GenBank mRNA |
AK098640.1 |
- |
3, 4 |
379 |
|
AL136651.1 |
- |
3, 4 |
339 |
|
BC136764.1 |
- |
3, 4 |
362 |
9020754 |
BC136765.1 |
- |
3, 4 |
362 |
9020755 |
BI550796.1 |
- |
3, 4 |
285 |
|
CR936752.1 |
- |
3, 4 |
344 |
|
CR936754.1 |
- |
3, 4 |
354 |
|
CR936764.1 |
- |
3, 4 |
341 |
|
View Full Product Details