Gene Transcripts

Gene Symbol
CARS2
Entrez Gene ID
79587
Gene Name
cysteinyl-tRNA synthetase 2, mitochondrial (putative)
Gene Aliases
COXPD27, cysRS
Chromosome Location
Chr. 13: 110641410 - 110713527 on Build GRCh38
UniGene ID
Hs.508725
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024537.3 NP_078813.1 3 413
XM_006719953.2 XP_006720016.1 3 493
XM_011521116.1 XP_011519418.1 3 205
XM_011521117.2 XP_011519419.1 3 349
XM_011521118.2 XP_011519420.1 3 349
XM_017020741.1 XP_016876230.1 2 141
XM_017020742.1 XP_016876231.1 3 349
GenBank mRNA AB062436.1 - 3 324
AK022180.1 - 3 349
AK301907.1 - 2 518
BC007220.1 - 3 336 2822557
DA966145.1 - 3 368
HY150557.1 - 3 414
HY160154.1 - 3 395

View Full Product Details