Gene Transcripts

Gene Symbol
FAAP100
Entrez Gene ID
80233
Gene Name
Fanconi anemia core complex associated protein 100
Gene Aliases
C17orf70
Chromosome Location
Chr. 17: 81539885 - 81552403 on Build GRCh38
UniGene ID
Hs.313905
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025161.5 NP_079437.5 9 3097
NR_033338.1 9 3293
XM_006722111.2 XP_006722174.1 9 2678
XM_006722112.3 XP_006722175.1 9 3243
XM_017025162.1 XP_016880651.1 9 3118
XM_017025163.1 XP_016880652.1 9 3014
GenBank mRNA AK024644.1 - 8 1524
AK025828.1 - 5 1358
AK054713.1 - 5 2441
BC001724.1 - 4 856
BC008883.2 - 9 3266
BC021968.2 - 5 1169
BC117139.1 - 7 2617 40125690
BC117141.1 - 7 2617 40125692
BC143861.1 - 7 2589
DQ989324.1 - 9 3079

View Full Product Details