Gene Transcripts

Gene Symbol
PRPS1
Entrez Gene ID
5631
Gene Name
phosphoribosyl pyrophosphate synthetase 1
Gene Aliases
ARTS, CMTX5, DFN2, DFNX1, PPRibP, PRS-I, PRSI
Chromosome Location
Chr. X: 107628424 - 107651026 on Build GRCh38
UniGene ID
Hs.56
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204402.1 NP_001191331.1 4 1171
NM_002764.3 NP_002755.1 7 1579
GenBank mRNA AK223169.1 - 7 1396
AK307748.1 - 6 1395
AK316467.1 - 4 1085
BC001605.1 - 7 1474 3542584
D00860.1 - 7 1496

View Full Product Details