Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001011663.1 | NP_001011663.1 | 1 | 322 | |
NM_032154.3 | NP_115530.2 | 1 | 322 | ||
GenBank mRNA | AB047006.1 | - | 1 | 276 | |
AK027885.1 | - | 1 | 276 | ||
AK290679.1 | - | 1 | 276 | ||
AK309105.1 | - | 1 | 344 | ||
BC007602.1 | - | 1 | 273 | 3350074 | |
BC010235.1 | - | 1 | 328 | 3461513 |