Gene Transcripts
- Gene Symbol
- C22orf23
- Entrez Gene ID
- 84645
- Gene Name
- chromosome 22 open reading frame 23
- Gene Aliases
- EVG1, dJ1039K5.6
- Chromosome Location
- Chr. 22: 37943050 - 37953669 on Build GRCh38
- UniGene ID
- Hs.517612
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001207062.1 |
NP_001193991.1
|
3, 4 |
356 |
|
NM_032561.4 |
NP_115950.3
|
4, 5 |
567 |
|
XM_005261781.1 |
XP_005261838.1
|
4, 5 |
531 |
|
XM_005261782.3 |
XP_005261839.1
|
4, 5 |
411 |
|
XM_005261783.2 |
XP_005261840.1
|
3, 4 |
468 |
|
XM_005261784.2 |
XP_005261841.1
|
3, 4 |
468 |
|
GenBank mRNA |
AF324466.1 |
- |
3, 4 |
367 |
|
AK057349.1 |
- |
4, 5 |
617 |
|
BC031998.1 |
- |
4, 5 |
449 |
4755328 |
BI463610.1 |
- |
4, 5 |
572 |
|
BI827388.1 |
- |
3, 4 |
356 |
|
BP368870.1 |
- |
3, 4 |
450 |
|
CR456440.1 |
- |
4, 5 |
447 |
|
View Full Product Details