Gene Transcripts
- Gene Symbol
- CELF6
- Entrez Gene ID
- 60677
- Gene Name
- CUGBP, Elav-like family member 6
- Gene Aliases
- BRUNOL6
- Chromosome Location
- Chr. 15: 72284727 - 72320184 on Build GRCh38
- UniGene ID
- Hs.348342
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001172684.1 |
NP_001166155.1
|
3, 4 |
476 |
|
NM_001172685.1 |
NP_001166156.1
|
2, 3 |
234 |
|
NM_052840.4 |
NP_443072.3
|
3, 4 |
714 |
|
GenBank mRNA |
AF401233.1 |
- |
3, 4 |
659 |
|
AF425606.1 |
- |
3, 4 |
475 |
|
AK124858.1 |
- |
2, 3 |
268 |
|
AK131098.1 |
- |
5, 6 |
1863 |
|
AK294382.1 |
- |
2, 3 |
234 |
|
AK296004.1 |
- |
3, 4 |
412 |
|
BC030835.1 |
- |
3, 4 |
417 |
4793742 |
BC033838.2 |
- |
3, 4 |
318 |
4214670 |
DC368992.1 |
- |
3, 4 |
475 |
|
View Full Product Details