Gene Transcripts
- Gene Symbol
- MBD6
- Entrez Gene ID
- 114785
- Gene Name
- methyl-CpG binding domain protein 6
- Gene Aliases
- Chromosome Location
- Chr. 12: 57520710 - 57530148 on Build GRCh38
- UniGene ID
- Hs.524523
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_052897.3 |
NP_443129.3
|
4, 5 |
448 |
|
XM_005268617.1 |
XP_005268674.1
|
4, 5 |
501 |
|
XM_005268618.1 |
XP_005268675.1
|
4, 5 |
597 |
|
XM_005268620.2 |
XP_005268677.1
|
4, 5 |
501 |
|
XM_006719217.1 |
XP_006719280.1
|
4, 5 |
346 |
|
XM_006719218.1 |
XP_006719281.1
|
4, 5 |
334 |
|
XM_011537851.2 |
XP_011536153.1
|
4, 5 |
1334 |
|
GenBank mRNA |
AB067474.1 |
- |
1, 2 |
117 |
|
AK056399.1 |
- |
4, 5 |
482 |
|
BC065530.1 |
- |
4, 5 |
448 |
6152748 |
View Full Product Details