Gene Transcripts

Gene Symbol
ZNF830
Entrez Gene ID
91603
Gene Name
zinc finger protein 830
Gene Aliases
CCDC16, OMCG1
Chromosome Location
Chr. 17: 34961530 - 34963186 on Build GRCh38
UniGene ID
Hs.413678
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_052857.3 NP_443089.3 1 552
GenBank mRNA AK055707.1 - 1 536
AK315671.1 - 1 552
BC002913.2 - 1 497
BC009044.1 - 1 115
BC011584.1 - 1 497
DB030670.1 - 1 552

View Full Product Details