Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001079536.1 | NP_001073004.1 | 5 | 1217 | |
NM_001297668.1 | NP_001284597.1 | 4 | 1452 | ||
NM_053017.4 | NP_443750.2 | 4 | 1530 | ||
NR_123732.1 | 4 | 582 | |||
XM_005252779.3 | XP_005252836.1 | 4 | 1332 | ||
XM_006718136.3 | XP_006718199.1 | 5 | 1230 | ||
XM_011519879.2 | XP_011518181.1 | 5 | 1380 | ||
XM_017017171.1 | XP_016872660.1 | 5 | 846 | ||
GenBank mRNA | AY358466.1 | - | 4 | 1077 | |
BC014577.1 | - | 4 | 1221 | 3934249 | |
Y16835.1 | - | 5 | 1220 |