Gene Transcripts

Gene Symbol
ASB12
Entrez Gene ID
142689
Gene Name
ankyrin repeat and SOCS box containing 12
Gene Aliases
Chromosome Location
Chr. X: 64224192 - 64230631 on Build GRCh38
UniGene ID
Hs.56281
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_130388.3 NP_569059.3 2 303
GenBank mRNA AF403030.1 - 1 83
AK096896.1 - 2 283
AK300684.1 - 2 279
BC069436.1 - 1 83 7262244
BC069555.1 - 1 83 7262256
BC093883.1 - 1 144 7939728
BC104946.1 - 1 144 8143949
DA560184.1 - 2 305

View Full Product Details