Gene Transcripts

Gene Symbol
TOR2A
Entrez Gene ID
27433
Gene Name
torsin family 2 member A
Gene Aliases
TORP1
Chromosome Location
Chr. 9: 127731524 - 127735387 on Build GRCh38
UniGene ID
Hs.444106
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001085347.2 NP_001078816.1 2, 3 507
NM_001134430.2 NP_001127902.1 2, 3 507
NM_001134431.2 NP_001127903.1 1, 2 241
NM_001252018.1 NP_001238947.1 1, 2 241
NM_001252023.1 NP_001238952.1 1, 2 241
NM_130459.3 NP_569726.2 2, 3 507
XM_011518554.1 XP_011516856.1 2, 3 542
GenBank mRNA AK027677.1 - 2, 3 442
AK055034.1 - 1, 2 186
AK075520.1 - 2, 3 450
AK092388.1 - 1 1758
AY358180.1 - 2, 3 474
BC062340.1 - 1, 2 206
BC080527.1 - 1, 2 167
BC100907.1 - 1, 2 176
BC100908.2 - 1, 2 176
BC100909.2 - 1, 2 171
BC100910.1 - 1, 2 176
BM695589.1 - 1, 2 182
BM772287.1 - 1, 2 184
CN263138.1 - 2, 3 509
DA430836.1 - 2, 3 483

View Full Product Details