Gene Transcripts

Gene Symbol
CMTM5
Entrez Gene ID
116173
Gene Name
CKLF like MARVEL transmembrane domain containing 5
Gene Aliases
CKLFSF5
Chromosome Location
Chr. 14: 23376439 - 23379772 on Build GRCh38
UniGene ID
Hs.99272
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037288.1 NP_001032365.1 4 1000
NM_001288744.1 NP_001275673.1 4 940
NM_001288745.1 NP_001275674.1 3 847
NM_001288746.1 NP_001275675.1 6 1294
NM_138460.2 NP_612469.1 5 1093
XM_017020953.1 XP_016876442.1 6 1079
XM_017020954.1 XP_016876443.1 6 956
GenBank mRNA AF479262.2 - 5 1094
AF527413.1 - 6 1077
AF527414.1 - 5 724
AF527948.2 - 3 618
AF527949.1 - 4 711
AK094840.1 - 4 1825
BC013109.1 - 5 838 4214683

View Full Product Details