Gene Transcripts
- Gene Symbol
- RHOT2
- Entrez Gene ID
- 89941
- Gene Name
- ras homolog family member T2
- Gene Aliases
- ARHT2, C16orf39, MIRO-2, MIRO2, RASL
- Chromosome Location
- Chr. 16: 668083 - 674174 on Build GRCh38
- UniGene ID
- Hs.513242
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_138769.2 |
NP_620124.1
|
4, 5 |
335 |
|
XM_005255660.2 |
XP_005255717.1
|
4, 5 |
286 |
|
XM_005255663.2 |
XP_005255720.1
|
4, 5 |
286 |
|
XM_006720970.2 |
XP_006721033.1
|
4, 5 |
286 |
|
XM_006720973.2 |
XP_006721036.1
|
4, 5 |
286 |
|
XM_017023825.1 |
XP_016879314.1
|
4, 5 |
286 |
|
XM_017023826.1 |
XP_016879315.1
|
4, 5 |
286 |
|
XM_017023829.1 |
XP_016879318.1
|
4, 5 |
286 |
|
XM_017023831.1 |
XP_016879320.1
|
4, 5 |
286 |
|
XM_017023833.1 |
XP_016879322.1
|
4, 5 |
286 |
|
XM_017023835.1 |
XP_016879324.1
|
4, 5 |
284 |
|
XM_017023836.1 |
XP_016879325.1
|
4, 5 |
286 |
|
XM_017023837.1 |
XP_016879326.1
|
4, 5 |
286 |
|
XM_017023839.1 |
XP_016879328.1
|
4, 5 |
284 |
|
XM_017023840.1 |
XP_016879329.1
|
4, 5 |
216 |
|
XM_017023841.1 |
XP_016879330.1
|
4, 5 |
275 |
|
XM_017023842.1 |
XP_016879331.1
|
4, 5 |
276 |
|
GenBank mRNA |
AJ517413.1 |
- |
4, 5 |
218 |
|
AK090426.1 |
- |
1, 2 |
38 |
|
AL834163.1 |
- |
3, 4 |
369 |
|
AY207375.1 |
- |
4, 5 |
332 |
|
BC014942.1 |
- |
4, 5 |
285 |
4859240 |
DB465261.1 |
- |
4, 5 |
336 |
|
View Full Product Details