Gene Transcripts
- Gene Symbol
- A2ML1
- Entrez Gene ID
- 144568
- Gene Name
- alpha-2-macroglobulin like 1
- Gene Aliases
- CPAMD9
- Chromosome Location
- Chr. 12: 8822472 - 8887202 on Build GRCh38
- UniGene ID
- Hs.620532
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001282424.2 |
NP_001269353.1
|
21, 22 |
2840 |
|
NM_144670.5 |
NP_653271.2
|
32, 33 |
4325 |
|
XM_011520566.2 |
XP_011518868.1
|
32, 33 |
4282 |
|
XM_011520567.2 |
XP_011518869.1
|
32, 33 |
4282 |
|
XM_017018868.1 |
XP_016874357.1
|
32, 33 |
4279 |
|
XM_017018869.1 |
XP_016874358.1
|
32, 33 |
4279 |
|
XM_017018870.1 |
XP_016874359.1
|
32, 33 |
4243 |
|
GenBank mRNA |
AK057908.1 |
- |
3, 4 |
327 |
|
AK096448.1 |
- |
10, 11 |
1314 |
|
AK122624.1 |
- |
26, 27 |
3411 |
|
AK123591.1 |
- |
15, 16 |
1978 |
|
AK123592.1 |
- |
21, 22 |
2795 |
|
AK123601.1 |
- |
29, 30 |
3702 |
|
AK290355.1 |
- |
32, 33 |
4178 |
|
AK302555.1 |
- |
21, 22 |
2840 |
|
AL832139.1 |
- |
32, 33 |
4176 |
|
AL832750.1 |
- |
10, 11 |
1050 |
|
AL833479.1 |
- |
30, 31 |
3841 |
|
BC093840.1 |
- |
3, 4 |
278 |
|
BC112131.1 |
- |
3, 4 |
278 |
|
BX647674.1 |
- |
14, 15 |
1780 |
|
BX648231.1 |
- |
33, 34 |
4177 |
|
View Full Product Details