Gene Transcripts
- Gene Symbol
- C11orf40
- Entrez Gene ID
- 143501
- Gene Name
- chromosome 11 open reading frame 40
- Gene Aliases
- NOV1
- Chromosome Location
- Chr. 11: 4571423 - 4577820 on Build GRCh38
- UniGene ID
- Hs.350556
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_144663.1 |
NP_653264.1
|
3, 4 |
450 |
|
GenBank mRNA |
AF439154.1 |
- |
3, 4 |
450 |
|
View Full Product Details