Gene Transcripts
- Gene Symbol
- PROSER3
- Entrez Gene ID
- 148137
- Gene Name
- proline and serine rich 3
- Gene Aliases
- C19orf55
- Chromosome Location
- Chr. 19: 35758139 - 35770115 on Build GRCh38
- UniGene ID
- Hs.527982
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001039887.2 |
NP_001034976.2
|
5, 6 |
612 |
|
XM_011526528.1 |
XP_011524830.1
|
5, 6 |
602 |
|
XM_011526529.2 |
XP_011524831.1
|
5, 6 |
613 |
|
XM_011526530.1 |
XP_011524832.1
|
5, 6 |
602 |
|
XM_011526531.1 |
XP_011524833.1
|
5, 6 |
602 |
|
XM_011526532.1 |
XP_011524834.1
|
5, 6 |
602 |
|
XM_011526533.2 |
XP_011524835.1
|
5, 6 |
602 |
|
XM_011526534.1 |
XP_011524836.1
|
5, 6 |
602 |
|
XM_011526535.2 |
XP_011524837.1
|
3, 4 |
309 |
|
XM_011526536.2 |
XP_011524838.1
|
5, 6 |
613 |
|
GenBank mRNA |
AK055219.1 |
- |
5, 6 |
568 |
|
BC110893.1 |
- |
4, 5 |
543 |
3914279 |
View Full Product Details