Gene Transcripts

Gene Symbol
CIART
Entrez Gene ID
148523
Gene Name
circadian associated repressor of transcription
Gene Aliases
C1orf51, CHRONO, GM129
Chromosome Location
Chr. 1: 150282533 - 150287093 on Build GRCh38
UniGene ID
Hs.54680
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001300838.1 NP_001287767.1 5 710
NM_001300839.1 NP_001287768.1 5 456
NM_001300840.1 NP_001287769.1 5 404
NM_001300841.1 NP_001287770.1 5 472
NM_144697.3 NP_653298.1 4 1299
XM_017000378.1 XP_016855867.1 4 565
GenBank mRNA AK098755.1 - 3 907
AK315397.1 - 4 984
BC017397.1 - 5 428
BC027999.1 - 5 700 5194413
BC047238.1 - 4 1013 5273646
BE784098.1 - 5 402
CN359562.1 - 5 370

View Full Product Details