Gene Transcripts
- Gene Symbol
- DDIAS
- Entrez Gene ID
- 220042
- Gene Name
- DNA damage induced apoptosis suppressor
- Gene Aliases
- C11orf82, noxin
- Chromosome Location
- Chr. 11: 82901695 - 82934659 on Build GRCh38
- UniGene ID
- Hs.165607
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_145018.3 |
NP_659455.3
|
3, 4 |
365 |
|
XM_011544835.1 |
XP_011543137.1
|
2, 3 |
227 |
|
XM_011544836.2 |
XP_011543138.1
|
2, 3 |
182 |
|
XM_011544838.2 |
XP_011543140.1
|
3, 4 |
329 |
|
GenBank mRNA |
AK058145.1 |
- |
4, 5 |
576 |
|
AK096157.1 |
- |
3, 4 |
330 |
|
AK292399.1 |
- |
3, 4 |
324 |
|
AK299581.1 |
- |
3, 4 |
323 |
|
BC039268.1 |
- |
3, 4 |
333 |
5296465 |
DB454086.1 |
- |
3, 4 |
368 |
|
View Full Product Details