Gene Transcripts

Gene Symbol
FRA10AC1
Entrez Gene ID
118924
Gene Name
fragile site, folic acid type, rare, fra(10)(q23.3) or fra(10)(q24.2) candidate 1
Gene Aliases
C10orf4, F26C11.1-like, FRA10A
Chromosome Location
Chr. 10: 93667883 - 93702572 on Build GRCh38
UniGene ID
Hs.586650
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145246.4 NP_660289.2 2, 3 283
XM_005269515.4 XP_005269572.1 3, 4 380
XM_005269516.4 XP_005269573.1 2, 3 475
XM_011539259.2 XP_011537561.1 2, 3 248
XM_011539260.2 XP_011537562.1 3, 4 338
XM_017015656.1 XP_016871145.1 2, 3 248
XM_017015657.1 XP_016871146.1 2, 3 283
XM_017015658.1 XP_016871147.1 3, 4 383
XM_017015659.1 XP_016871148.1 2, 3 475
XM_017015660.1 XP_016871149.1 3, 4 340
XM_017015661.1 XP_016871150.1 3, 4 397
XM_017015662.1 XP_016871151.1 2, 3 248
GenBank mRNA AK090955.1 - 2, 3 283
AK308861.1 - 2, 3 201
AK309016.1 - 2, 3 396
BM272089.1 - 2, 3 267
BX648699.1 - 1, 2 317
CN298116.1 - 2, 3 262

View Full Product Details