Gene Transcripts
- Gene Symbol
- TYW5
- Entrez Gene ID
- 129450
- Gene Name
- tRNA-yW synthesizing protein 5
- Gene Aliases
- C2orf60, hTYW5
- Chromosome Location
- Chr. 2: 199928911 - 199955736 on Build GRCh38
- UniGene ID
- Hs.204619
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001039693.2 |
NP_001034782.1
|
6, 7 |
848 |
|
NR_004862.1 |
|
6, 7 |
829 |
|
NR_109907.1 |
|
5, 6 |
693 |
|
XM_017003310.1 |
XP_016858799.1
|
4, 5 |
476 |
|
GenBank mRNA |
AK095272.1 |
- |
5, 6 |
457 |
|
AK298977.1 |
- |
6, 7 |
592 |
|
BC063502.1 |
- |
2, 3 |
117 |
|
BC132835.1 |
- |
6, 7 |
612 |
40146857 |
BC136837.1 |
- |
6, 7 |
612 |
9020828 |
BC144420.1 |
- |
6, 7 |
593 |
|
BX482572.1 |
- |
6, 7 |
602 |
|
BX647677.1 |
- |
6, 7 |
830 |
|
CN261486.1 |
- |
6, 7 |
561 |
|
View Full Product Details