Gene Transcripts

Gene Symbol
LRRC75A-AS1
Entrez Gene ID
125144
Gene Name
LRRC75A antisense RNA 1
Gene Aliases
C17orf45, C17orf76-AS1, FAM211A-AS1, NCRNA00188, TSAP19
Chromosome Location
Chr. 17: 16438987 - 16470648 on Build GRCh38
UniGene ID
Hs.368934
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027158.1 4 739
NR_027159.1 4 1062
NR_027160.1 5 1185
NR_027161.1 4 1181
NR_027162.1 5 1238
NR_027163.1 6 972
NR_027164.1 6 919
NR_027165.1 5 783
NR_027166.1 5 796
NR_027167.1 5 862
NR_027168.2 6 888
NR_027169.1 5 915
NR_027170.1 6 840
NR_027667.1 5 1106
NR_045021.1 5 775
NR_045022.1 6 914
NR_045027.1 3 1005
NR_045028.1 4 1049
NR_045029.1 5 870
GenBank mRNA AJ012499.1 - 1 293
AK098643.1 - 5 742
AK309961.1 - 5 812
BC012396.1 - 3 659
BC019833.1 - 4 721
BC027986.1 - 5 1057 4940349
BC040159.1 - 5 1031 5539041
BX538291.1 - 6 791
CD050014.1 - 5 767

View Full Product Details