Gene Transcripts

Gene Symbol
CDCA2
Entrez Gene ID
157313
Gene Name
cell division cycle associated 2
Gene Aliases
PPP1R81, Repo-Man
Chromosome Location
Chr. 8: 25458569 - 25511308 on Build GRCh38
UniGene ID
Hs.33366
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317906.1 NP_001304835.1 14 3328
NM_001317907.1 NP_001304836.1 15 3264
NM_152562.3 NP_689775.2 15 3245
XM_006716294.2 XP_006716357.1 15 3260
XM_011544415.1 XP_011542717.1 16 3332
XM_011544416.2 XP_011542718.1 16 3026
XM_011544417.2 XP_011542719.1 16 3083
XM_011544418.1 XP_011542720.1 16 3296
GenBank mRNA AK092164.1 - 10 1861
AK098670.1 - 2 1022
AK292505.1 - 14 3060
AL833396.1 - 12 2350
AL833627.1 - 14 3022
BC036214.1 - 12 2373 5267276
BC063651.1 - 2 953
BC085609.1 - 2 1008
BG354575.1 - 2 1029
BX649008.1 - 11 2236

View Full Product Details