Gene Transcripts

Gene Symbol
AKNAD1
Entrez Gene ID
254268
Gene Name
AKNA domain containing 1
Gene Aliases
C1orf62
Chromosome Location
Chr. 1: 108815898 - 108858242 on Build GRCh38
UniGene ID
Hs.729441
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152763.4 NP_689976.2 2 401
NR_049760.1 3 542
GenBank mRNA AK095517.1 - 3 528
AK313772.1 - 2 381
AL832216.1 - 2 391
BC030647.2 - 2 381
BC150642.1 - 1 156 9057012
DB054557.1 - 2 402

View Full Product Details