Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001321395.1 | NP_001308324.1 | 3 | 439 | |
NM_152781.3 | NP_689994.2 | 3 | 439 | ||
NR_135630.1 | 3 | 439 | |||
XM_005278233.2 | XP_005278290.1 | 4 | 398 | ||
XM_005278235.1 | XP_005278292.1 | 4 | 403 | ||
XM_006721802.3 | XP_006721865.1 | 1 | 54 | ||
XM_011524586.2 | XP_011522888.1 | 3 | 382 | ||
XM_017024424.1 | XP_016879913.1 | 3 | 419 | ||
GenBank mRNA | AK057392.1 | - | 3 | 439 | |
AK301774.1 | - | 4 | 403 | ||
AK301884.1 | - | 4 | 403 | ||
AK301943.1 | - | 3 | 419 | ||
BC033734.1 | - | 3 | 387 | ||
BC132681.1 | - | 3 | 375 | 40146703 | |
BC144011.1 | - | 3 | 375 |