Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_152945.3 | NP_694453.2 | 2, 3 | 554 | |
XM_005246287.4 | XP_005246344.1 | 2, 3 | 564 | ||
XM_017003320.1 | XP_016858809.1 | 2, 3 | 997 | ||
XM_017003321.1 | XP_016858810.1 | 2, 3 | 998 | ||
XM_017003322.1 | XP_016858811.1 | 2, 3 | 563 | ||
GenBank mRNA | AB036991.1 | - | 2, 3 | 476 | |
AF526533.1 | - | 2, 3 | 554 | ||
AK126575.1 | - | 2, 3 | 412 | ||
BC066549.1 | - | 2, 3 | 523 | 5748065 |