Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_153018.2 | NP_694563.1 | 2 | 347 | |
GenBank mRNA | AK055288.1 | - | 2 | 347 | |
AK290549.1 | - | 2 | 339 | ||
BC142962.1 | - | 2 | 326 | 8860374 | |
BP250703.1 | - | 2 | 347 | ||
BX647638.1 | - | 2 | 300 |